missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Ab Buffer Kit
Description
Includes
Binding buffer (10X stock solution), 50mL; Elution buffer (10X stock solution), 15mL; Neutralizing buffer, 25mL
- Eliminates time-consuming buffer preparation
- Optimized protocols for Ab SpinTrap™, Protein A HP SpinTrap, Protein G HP SpinTrap columns and HiTrap™ columns prepacked with either Protein A or Protein G media
Specifications
Specifications
| Chemical Name or Material | Ab Buffer kit |
| For Use With (Application) | Used for binding, washing, elution of IgG according to recommended protocols, purification of Monoclonal and Polyclonal IgG using immobilized Protein A or Protein G |
| For Use With (Equipment) | Ab SpinTrap™, Protein A HP SpinTrap, Protein G HP SpinTrap™ columns and HiTrap™ columns |
| Quantity | Each |
| Includes | 50mL Binding Buffer (10X stock solution), 15mL Elution Buffer (10X stock solution), 25mL Neutralizing Buffer |