Learn More
ACROBiosystems Inc Monkeypox virus (strain Zaire-96-I-16) A35R , His Tag (A3R-M52H3) is expressed from human 293 cells (HEK293). It contains AA Arg 58 - Thr 181 (Accession # Q8V4U4).
Catalog No. 502253380
Encompass
Monkeypox is a rare zoonosis caused by monkeypox virus, which has become the most serious orthpoxvirus and consists of complex double stranded DNA. The cases are mostly in central and western Africa. The pathogenesis of monkeypox is that the virus invades the body from respiratory mucosa, multiplies in lymphocytes, and incurs into blood producing transient venereal toxemia. after the virus multiplies in cells, the cells can invade the blood and propagate to the skin of the whole body, causing lesions. The envelope glycoprotein A35R on the EV surface has been predicted to influence intercellular diffusion of virions.
Catalog No. 502253380 Supplier ACROBiosystems Inc Supplier No. A3RM52H3100UG
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Monkeypox is a rare zoonosis caused by monkeypox virus, which has become the most serious orthpoxvirus and consists of complex double stranded DNA. The cases are mostly in central and western Africa. The pathogenesis of monkeypox is that the virus invades the body from respiratory mucosa, multiplies in lymphocytes, and incurs into blood producing transient venereal toxemia. after the virus multiplies in cells, the cells can invade the blood and propagate to the skin of the whole body, causing lesions. The envelope glycoprotein A35R on the EV surface has been predicted to influence intercellular diffusion of virions.