missing translation for 'onlineSavingsMsg'
Learn More

Cupric Nitrate, Hydrate, Crystal, Purified, Spectrum™ Chemical

Catalog No. 18601911
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
45 kg
Packaging:
Poly Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18601911 45 kg Poly Bottle
18601907 125 g Poly Bottle
18601908 500 g Poly Bottle
18601909 2.5 kg Poly Bottle
18601910 12 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18601911 Supplier Spectrum Chemical Mfg Cor Supplier No. C140745KGBL
Get Up to $500 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

C1407, 19004-19-4, Cu(NO3)2·2.5H2O

Spectrum™ Chemical Cupric Nitrate, Hydrate, Crystal, Purified is a also known as a copper nitrate and can be obtained as a crystalline solid as well as a hydrated form. Mostly used to produce copper oxide, it is also used as a polisher for other metals and in pyrotechnics.

Chemical Identifiers

CAS 19004-19-4
Molecular Formula Cu2H10N4O17
Molecular Weight (g/mol) 465.18
InChI Key IZDJJEMZQZQQQQ-UHFFFAOYSA-N
IUPAC Name Dicopper(2+) pentahydrate tetranitrate
SMILES O.O.O.O.O.[Cu++].[Cu++].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

CAS Max % 100%
Quantity 45 kg
Packaging Poly Bottle
UN Number UN1477
Grade Purified
Chemical Name or Material Cupric Nitrate