Learn More
ACROBiosystems Inc Monkeypox virus (strain Zaire-96-I-16) A29L , His Tag (A2L-M52H3) is expressed from human 293 cells (HEK293). It contains AA Met 1 - Glu 110 (Accession # Q77HM6).
Catalog No. 502253378
Encompass
Monkeypox is a rare zoonosis caused by monkeypox virus, which has become the most serious orthpoxvirus and consists of complex double stranded DNA. The cases are mostly in central and western Africa. The pathogenesis of monkeypox is that the virus invades the body from respiratory mucosa, multiplies in lymphocytes, and incurs into blood producing transient venereal toxemia. after the virus multiplies in cells, the cells can invade the blood and propagate to the skin of the whole body, causing lesions. A29L binds to cell surface heparin to promote fusion of viral membrane with host plasma membrane.
Catalog No. 502253378 Supplier ACROBiosystems Inc Supplier No. A2LM52H3100UG
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Monkeypox is a rare zoonosis caused by monkeypox virus, which has become the most serious orthpoxvirus and consists of complex double stranded DNA. The cases are mostly in central and western Africa. The pathogenesis of monkeypox is that the virus invades the body from respiratory mucosa, multiplies in lymphocytes, and incurs into blood producing transient venereal toxemia. after the virus multiplies in cells, the cells can invade the blood and propagate to the skin of the whole body, causing lesions. A29L binds to cell surface heparin to promote fusion of viral membrane with host plasma membrane.