Learn More
PTS Diagnostics A1CNow+™ Systems
Catalog No. 21124414
Encompass
Change view
Click to view available options
Quantity:
20-Count Test Kit
Catalog No. Quantity
21124414 20-Count Test Kit
1 options
Catalog No. 21124414 Supplier PTS Diagnostics Supplier No. 3021
Only null left
Provides fast and easy way of obtaining accurate A1C results
  • Results in five minutes
  • Requires only small volume (5μL) of blood sample
  • CLIA waived
  • Portable and can be used in multiple exam rooms
  • Easy to use and hence requires minimal training
  • Maintenance-free
  • Reimbursable
Certifications
National Glycohemoglobin Standardization Program (NGSP), IFCC
Specifications
Sample Type Blood
Quantity 20-Count Test Kit