Learn More
Enzo alpha-Synuclein (human) monoclonal antibody (5C2) (100l)
Catalog No. 502009966
Encompass
Clone: 5C2. Isotype: IgG1. Immunogen: Recombinant human -synuclein (aa 1-140). Species reactivity: Human. Specificity: Recognizes the NAC domain (aa 61-95) of human -synuclein. Applications: ELISA, WB. Purity: Protein G-affinity purified. Formulation: Liquid. In PBS, pH 7.4, containing 0.02% sodium azide and 10% glycerol.
Catalog No. 502009966 Supplier Enzo Supplier No. ALX804656R100
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Clone: 5C2. Isotype: IgG1. Immunogen: Recombinant human -synuclein (aa 1-140). Species reactivity: Human. Specificity: Recognizes the NAC domain (aa 61-95) of human -synuclein. Applications: ELISA, WB. Purity: Protein G-affinity purified. Formulation: Liquid. In PBS, pH 7.4, containing 0.02% sodium azide and 10% glycerol.