missing translation for 'onlineSavingsMsg'
Learn More
Learn More
A-803467, 98%, Thermo Scientific Chemicals
$144.04 - $910.90
Chemical Identifiers
| CAS | 944261-79-4 |
|---|---|
| Molecular Formula | C19H16ClNO4 |
| Molecular Weight (g/mol) | 357.79 |
| MDL Number | MFCD10574689 |
| InChI Key | VHKBTPQDHDSBSP-UHFFFAOYSA-N |
| PubChem CID | 16038374 |
| IUPAC Name | 5-(4-chlorophenyl)-N-(3,5-dimethoxyphenyl)furan-2-carboxamide |
| SMILES | COC1=CC(OC)=CC(NC(=O)C2=CC=C(O2)C2=CC=C(Cl)C=C2)=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAJ67281LB0
|
Thermo Scientific Chemicals
J67281LB0 |
5 mg |
Each for $144.04
|
|
|||||
|
AAJ67281MF
|
Thermo Scientific Chemicals
J67281MF |
50 mg |
Each for $910.90
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 944261-79-4 | |
| 357.79 | |
| VHKBTPQDHDSBSP-UHFFFAOYSA-N | |
| 5-(4-chlorophenyl)-N-(3,5-dimethoxyphenyl)furan-2-carboxamide |
| C19H16ClNO4 | |
| MFCD10574689 | |
| 16038374 | |
| COC1=CC(OC)=CC(NC(=O)C2=CC=C(O2)C2=CC=C(Cl)C=C2)=C1 |
Specifications
| 944261-79-4 | |
| White | |
| MFCD10574689 | |
| 5-4-chlorophenyl-n-3,5-dimethoxyphenyl furan-2-carboxamide, 5-4-chlorophenyl-n-3,5-dimethoxyphenyl-2-furancarboxamide, d0ff6s, 2-furancarboxamide, 5-4-chlorophenyl-n-3,5-dimethoxyphenyl, 5-4-chlorophenyl-n-3,5-dimethoxyphenyl, 5-4-chlorophenyl-furan-2-carboxylic acid 3,5, a hplc, 5-4-chlorophenyl-n-3,5-dimethoxyphenyl-2-furamide, 2-furancarboxamide,5-4-chlorophenyl-n-3,5-dimethoxyphenyl | |
| COC1=CC(OC)=CC(NC(=O)C2=CC=C(O2)C2=CC=C(Cl)C=C2)=C1 | |
| 357.79 | |
| 357.79 | |
| Solid |
| 128°C to 130°C | |
| C19H16ClNO4 | |
| 5 mg | |
| VHKBTPQDHDSBSP-UHFFFAOYSA-N | |
| 5-(4-chlorophenyl)-N-(3,5-dimethoxyphenyl)furan-2-carboxamide | |
| 16038374 | |
| 98% | |
| A-803467 |
Safety and Handling
P264b-P270-P301+P312-P330-P501c
H302
TSCA : No
storageNote1 : -30° to -10°C
Recommended Storage : Store at -20°C
RUO – Research Use Only