missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-6-methyl-2-methylthiopyrimidine, 97%, Thermo Scientific™

Catalog No. AC453750010
Change view
Click to view available options
Quantity:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC453750010 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC453750010 Supplier Thermo Scientific Chemicals Supplier No. 453750010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 17119-73-2
Molecular Formula C6H7ClN2S
Molecular Weight (g/mol) 174.65
InChI Key ALMBOXQFPLQVLF-UHFFFAOYSA-N
Synonym 4-chloro-6-methyl-2-methylthio pyrimidine, 4-chloro-6-methyl-2-methylsulfanyl pyrimidine, 4-chloro-6-methyl-2-methylsulfanyl-pyrimidine, pyrimidine, 4-chloro-6-methyl-2-methylthio, 4-chloro-6-methyl-2-methylthiopyrimidine, 2-methylmercapto-4-methyl-6-chloropyrimidine, pubchem21461, 6-chloro-4-methyl-2-methylthiopyrimidine, 4-chloro-6-methyl-2-methylthio-pyrimidine, 2-methylthio-4-methyl-6-chloropyrimidine
PubChem CID 241163
IUPAC Name 4-chloro-6-methyl-2-methylsulfanylpyrimidine
SMILES CC1=CC(=NC(=N1)SC)Cl

Specifications

Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1g
Formula Weight 174.65
Percent Purity 97%
Chemical Name or Material 4-Chloro-6-methyl-2-methylthiopyrimidine
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Harmful if swallowed. Causes skin irritation. Causes serious eye irritation. Harmful if inhaled. May cause respiratory irritation.
Precautionary Statement GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Call a POISON CENTER or doctor/physician if you feel unwell.

RUO – Research Use Only