missing translation for 'onlineSavingsMsg'
Learn More

3-Fluorobenzyl alcohol, 98%, Thermo Scientific™

Catalog No. AC171960050
Change view
Click to view available options
Quantity:
5g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC171960050 5g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC171960050 Supplier Thermo Scientific Chemicals Supplier No. 171960050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 456-47-3
Molecular Formula C7H7FO
Molecular Weight (g/mol) 126.13
MDL Number MFCD00004631
InChI Key QDHRSLFSDGCJFX-UHFFFAOYSA-N
Synonym 3-fluorobenzyl alcohol, 3-fluorophenyl methanol, m-fluorobenzyl alcohol, benzenemethanol, 3-fluoro, 3-fluorobenzylalcohol, 3-fluorobenzylic alcohol, benzyl alcohol, m-fluoro, 3-fluorophenyl methan-1-ol, 3-florobenzyl alcohol, pubchem3486
PubChem CID 68008
IUPAC Name (3-fluorophenyl)methanol
SMILES C1=CC(=CC(=C1)F)CO

Specifications

CAS Min % 97.5
CAS Max % 100.0
Density 1.1600g/mL
Boiling Point 104°C to 105°C (22.0mmHg)
Flash Point 90°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5080 to 1.511
Linear Formula FC6H4CH2OH
Quantity 5g
Specific Gravity 1.16
Formula Weight 126.13
Percent Purity 98%
Chemical Name or Material 3-Fluorobenzyl alcohol, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 207-265-3
TSCA TSCA

RUO – Research Use Only