Learn More
Sklar Instruments Scissors, Sklar klartMetzenbaum Dissecting Scissors, 9 in., Blunt/Blunt, Curved, Razor, Stainless Steel, EA.
Catalog No. 501093501
Encompass
Scissors, Sklar, Sklarlite Sklarhone Metzenbaum Dissecting Scissors, 9 in., Blunt/Blunt, Curved, Razor, Stainless Steel, EA. Sklarlite Sklarhone Metzenbaum Dissecting Scissors are used for dissecting and cutting tissue only. These scissors are not recommended for cutting sutures, drains, or heavy tissue. The Sklarlite Sklarhone combination provides the surgeon with a lightweight, precision instrument that allows the user to make the sharp incisions of a knife with the cutting ability of scissors, all while having greater control and experiencing less fatigue. This instrument is identified by dual black finger rings. This product is curved, has a smooth, razor edge with two blunt tips, and an overall length of 9 inches.
Catalog No. 501093501 Supplier Sklar Instruments Supplier No. 159119
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Scissors, Sklar, Sklarlite Sklarhone Metzenbaum Dissecting Scissors, 9 in., Blunt/Blunt, Curved, Razor, Stainless Steel, EA. Sklarlite Sklarhone Metzenbaum Dissecting Scissors are used for dissecting and cutting tissue only. These scissors are not recommended for cutting sutures, drains, or heavy tissue. The Sklarlite Sklarhone combination provides the surgeon with a lightweight, precision instrument that allows the user to make the sharp incisions of a knife with the cutting ability of scissors, all while having greater control and experiencing less fatigue. This instrument is identified by dual black finger rings. This product is curved, has a smooth, razor edge with two blunt tips, and an overall length of 9 inches.