missing translation for 'onlineSavingsMsg'
Learn More

9H-Pyrido[3,4-b]indole 98.0+%, TCI America™

Catalog No. P1121100MG
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P1121100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P1121100MG Supplier TCI America Supplier No. P1121100MG
Only null left

Chemical Identifiers

CAS 244-63-3
Molecular Formula C11H8N2
Molecular Weight (g/mol) 168.20
MDL Number MFCD00004956
InChI Key AIFRHYZBTHREPW-UHFFFAOYSA-N
Synonym 9h-pyrido 3,4-b indole, norharman, norharmane, beta-carboline, carbazoline, 2,9-diazafluorene, 9h-beta-carboline, 2-azacarbazole, .beta.-carboline
PubChem CID 64961
ChEBI CHEBI:109895
IUPAC Name 9H-pyrido[3,4-b]indole
SMILES N1C2=C(C=CC=C2)C2=C1C=NC=C2

Specifications

Melting Point 201°C
Color White-Yellow
Quantity 100 mg
Formula Weight 168.20
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 9H-Pyrido[3,4-b]indole
RTECSNumber UU9350000
TSCA No