Learn More
Thomson Instrument Company 96-Well Plate, 2mL, Square Well, Round Bottom, Irreversible, Sterile | CS20
Catalog No. NC1562847
Encompass
Thomson Instrument Company Well Plates are offered in 24 and 96-well configurations with V, pyramid, or U bottom, and square or round wells for cell culture and analytical sample preparation. Well plates are designed to SLAS specifications and work well on automated liquid handling systems. Plates are individually wrapped for your convince, and ideally designed to work with Thomson filter plates. Thomson manufactures its products in the USA to assure same day shipping.
Catalog No. NC1562847 Supplier Thomson Instrument Company Supplier No. 931130S
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Thomson Instrument Company Well Plates are offered in 24 and 96-well configurations with V, pyramid, or U bottom, and square or round wells for cell culture and analytical sample preparation. Well plates are designed to SLAS specifications and work well on automated liquid handling systems. Plates are individually wrapped for your convince, and ideally designed to work with Thomson filter plates. Thomson manufactures its products in the USA to assure same day shipping.