Learn More
ACROBiosystems Inc - 96tests
Catalog No. 502535127
Encompass
This ELISA Buffer Set-96T contains a High-bind 96well plate coating buffer washing buffer blocking buffer substrate solution and stop solution. The ELISA Buffer Set-96T can be quickly applied to the ELISA experiment and save your time to prepare buffers. This ELISA Buffer Set-96T is sterile-filtered.
Catalog No. 502535127 Supplier ACROBiosystems Inc Supplier No. EBS00296TESTS
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
This ELISA Buffer Set-96T contains a High-bind 96well plate coating buffer washing buffer blocking buffer substrate solution and stop solution. The ELISA Buffer Set-96T can be quickly applied to the ELISA experiment and save your time to prepare buffers. This ELISA Buffer Set-96T is sterile-filtered.