Learn More
Cytiva 96 Well Uniplates
Catalog No. PF77013250
Change view
Click to view available options
Color:
White
Catalog No. Color
PF77013250 White
1 options
Catalog No. PF77013250 Supplier Cytiva Supplier No. 77013250
Only null left
The 96 and 384 well format UNIPLATE with 'V' bottom is well suited for applications with small sample volumes
The 96 and 384 well format UNIPLATE with 'V' bottom is well suited for applications with small sample volumes. The vertical sides of the well, combined with the 'V' design at the base of each well, ensure that all the material runs down the side walls and is channeled into the well base. The 'V' bottom therefore ensures maximum sample recovery - typically approximately 99% liquid sample recovery is attained.
Specifications
Format Standard
Description GE’s Whatman UNIPLATE features a pointed bottom for high efficiency recovery of valuable, small volume samples. The vertical sides of the well ensure that material runs into the well base.
Array 8 x 12
Color White
Product Type Microplate
Material Polystyrene
No. of Wells 96
Total Volume (Metric) Well 250 μL
Well Shape Conical Bottom (V-well)
No. per Pack 50
Quantity 50/Pk.
Show More Show Less