missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eppendorf™ twin.tec™ PCR Plates LoBind™ - PCR Plates
Description
- LoBind material guarantees maximum sample recovery for improved assay results.
- One-piece design: combining a polycarbonate frame and polypropylene wells for optimum performance.
- Extremely thin-walled polypropylene wells guarantee optimum heat transfer to the sample.
- Exceptionally solid and torque-resistant polycarbonate frame.
- OptiTrack™ matrix for faster sample identification and fewer pipetting errors.
- Raised well rims for effective sealing, also reduces risk of cross-contamination.
- Batch-tested and certified free of DNA, DNAase, RNase and PCR inhibitors (PCR clean).
Specifications
Specifications
| Barcode | No Barcode |
| Certifications/Compliance | ISO 9001, ISO 13485, ISO 14001 |
| Format | Standard |
| Description | Clear — With optical guiding grid and laser-engraved lot number and expiration date |
| Array | 8 x 12 |
| Autoclavable | Autoclavable |
| Packaging | 5 Bags x 5 Plates |
| Product Line | twin.tec Trace |
| Color | Clear |
| Disposable | Single-use |
| Show More |