missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ twin.tec™ PCR Plates LoBind™ - PCR Plates

Catalog No. 14380052
Change view
Click to view available options
Color:
Clear
Green
No. of Wells:
384
96
Skirt Style:
Semi-Skirted
Skirted
Skirted, colorless
Packaging:
5 Bags x 5 Plates
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color No. of Wells Skirt Style Packaging
14380052 Clear 96 Skirted 5 Bags x 5 Plates
E0030129512 Clear 96 Skirted 5 Bags x 5 Plates
14380057 Clear 96 Skirted, colorless 5 Bags x 5 Plates
E0030129660 Green 96 Skirted 5 Bags x 5 Plates
14380054 Clear 96 Semi-Skirted 5 Bags x 5 Plates
E0030129504 Clear 96 Semi-Skirted 5 Bags x 5 Plates
14380053 Clear 384 Skirted 5 Bags x 5 Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 14380052 Supplier Eppendorf™ Supplier No. 0030531086
Only null left

Eppendorf twin.tec™ PCR plates LoBind, available in 96- and 384-well formats, maximize nucleic acid recovery and are certified as PCR clean for consistently reliable results.

  • LoBind material guarantees maximum sample recovery for improved assay results.
  • One-piece design: combining a polycarbonate frame and polypropylene wells for optimum performance.
  • Extremely thin-walled polypropylene wells guarantee optimum heat transfer to the sample.
  • Exceptionally solid and torque-resistant polycarbonate frame.
  • OptiTrack™ matrix for faster sample identification and fewer pipetting errors.
  • Raised well rims for effective sealing, also reduces risk of cross-contamination.
  • Batch-tested and certified free of DNA, DNAase, RNase and PCR inhibitors (PCR clean).

Specifications

Barcode No Barcode
Certifications/Compliance ISCC, ACT Label, ISO 9001, ISO 13485, ISO 14001
Format Standard
Description Clear — BioBased (made of bio-based material)
Array 8 x 12
Autoclavable Autoclavable
Packaging 5 Bags x 5 Plates
Product Line twin.tec Trace
Color Clear
Disposable Single-use
Product Type PCR Plate
Skirt Style Skirted
Material Polypropylene, Polycarbonate
Volume (Metric) Well 150 μL
No. of Wells 96
Sterility Non-sterile
Surface Treatment LoBind
Well Shape Conical Bottom (V-well)
Max. RCF 6,000 x g
Purity or Quality Grade PCR Clean (Non-sterile)
Quantity 25 Pk.
Rimmed Yes
Show More Show Less