Learn More
Celltreat Scientific Products 96 Well Non-treated Plate without Lid, Sterile
Catalog No. 50202150
Encompass
CELLTREAT Scientific Products 96 Well Non-Treated Plate without Lid. Tissue Culture Treated surface improves cell adhesion. Plates are labeled with alpha-numeric markings for easy identification of individual wells. Each well has a raised rim which nests into the rings on the single direction lid to reduce evaporation and limit the risk of cross contamination. Lids have a stacking feature for added stability when handling tall stacks of plates. Flat bottom with working volume of 50-300L. RNase/Dnase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Individually packed sterile, 100/case.
Catalog No. 50202150 Supplier Celltreat Scientific Products Supplier No. 229592
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CELLTREAT Scientific Products 96 Well Non-Treated Plate without Lid. Tissue Culture Treated surface improves cell adhesion. Plates are labeled with alpha-numeric markings for easy identification of individual wells. Each well has a raised rim which nests into the rings on the single direction lid to reduce evaporation and limit the risk of cross contamination. Lids have a stacking feature for added stability when handling tall stacks of plates. Flat bottom with working volume of 50-300L. RNase/Dnase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Individually packed sterile, 100/case.