missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ 96-well tips, 50 μL

Catalog No. 361041695
Change view
Click to view available options
Volume (Metric):
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
361041695 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 361041695 Supplier Axygen™ Supplier No. RRP50CR

Axygen™ offers a comprehensive line of tips for PerkinElmer™ automation systems.

These tips are manufactured to stringent specifications under strict process controls and assembled via automation to ensure consistent performance and quality. Tips are RNase-/DNase-free and nonpyrogenic. Available in non-sterile, sterile, non-filtered and filtered. Clear and Conductive Black polypropylene tips are available in 96-well format designed for PerkinElmer™ Janus™/MultiProbe Variospan-1, 4, 8 head.

  • 50 μL Clear Tips
  • 96 Tip format
  • Clear Polypropylene
  • RNase-/DNase-free
  • Nonpyrogenic
  • non-sterile
  • Compatibility: PerkinElmer™ Janus™/MultiProbe Variospan-1, 4, 8 head

Specifications

Volume (Metric) 50 μL
Filtered Non-filtered
Sterility Non-sterile
Color Clear
Format Rack
Tip Style Standard, Round
Packaging SLAS Rack
Quantity 96 Tips/Rk, 10 Racks/Pk., 5 Pk./Cs
For Use With (Equipment) PerkinElmer, Tecan