missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Abgene™ 96-well Sealing Mats for Sample Processing & Storage DeepWell™ and MicroWell Microplates
GREENER_CHOICE

Catalog No. AB0675
Change view
Click to view available options
Temperature Range (Metric):
-20°C to +130°C
-20°C to +55°C
Well Shape:
Round
Square
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Temperature Range (Metric) Well Shape
AB0675 -20°C to +130°C Square
AB0674 -20°C to +130°C Round
AB0566 -20°C to +55°C Round
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AB0675 Supplier Thermo Scientific™ Supplier No. AB0675

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Round or square CapMats, Sealing Mats, MatCaps for sealing 96-well storage plates, available in autoclavable and pierceable formats.

Thermo Scientific 96-well Sealing Mats are designed for 96-well storage plates and are available in either pierceable or autoclavable formats. The round or square capped mats are made of thermoplastic elastomer and are suitable for long-term storage.

Highlights

  • Certified RNAse, DNAse, PCR Inhibitor, and human DNA free
  • Round or square capped mats for sealing 96-well storage plates
  • Alphanumeric grid-referencing
  • Molded from a proprietary thermoplastic elastomer for a flexible design
  • Suitable for long term storage
  • Available in autoclavable or pierceable formats

Pierceable version:

  • Solvent resistant (including DMSO, Ethanol, and Isopropanol)
  • Suitable for Gamma Irradiation
  • Requires 6N for piercing
  • Temperature Range: -20°C to +55°C

Autoclavable version:

  • Suitable for Gamma Irradiation
  • Temperature Range: -20°C to +130°C

Hazardous: No

Storage Condition: Ambient

Specifications

Color Natural
Solvent Or Alcohol Resistant Alcohol Resistant, Solvent Resistant
Description CapMat for 96-well Polypropylene Sample Processing & Storage Plates
Certifications/Compliance Certified RNase, DNase, PCR Inhibitors, and human DNA Free. ANSI Compliant
Material TPE
Sterility Sterile
Autoclavable Autoclavable
No. of Wells 96
No. per Case 50
No. per Pack 10
Plug Diameter 8.4mm
Temperature Range (English) -4°F to 266°F
For Use With (Application) Designed for sealing 96-well storage plate and suitable for long term storage
Format 96-well
Product Type 96-well capmat, sealing mat
Product Line Abgene
Adhesive None
Certified RNase/DNase-free Yes
Pierceable No
Temperature Range (Metric) -20°C to +130°C
Well Shape Square
Plate Compatibility Abgene 96 Square Well
Fit for Purpose PCR Ready
Green Features ACT Ecolabel (Silver)
Show More Show Less