missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Abgene™ 96-well Sealing Mats for Sample Processing & Storage DeepWell™ and MicroWell Microplates

Description
Round or square CapMats, Sealing Mats, MatCaps for sealing 96-well storage plates, available in autoclavable and pierceable formats.
Thermo Scientific 96-well Sealing Mats are designed for 96-well storage plates and are available in either pierceable or autoclavable formats. The round or square capped mats are made of thermoplastic elastomer and are suitable for long-term storage.
Highlights
- Certified RNAse, DNAse, PCR Inhibitor, and human DNA free
- Round or square capped mats for sealing 96-well storage plates
- Alphanumeric grid-referencing
- Molded from a proprietary thermoplastic elastomer for a flexible design
- Suitable for long term storage
- Available in autoclavable or pierceable formats
Pierceable version:
- Solvent resistant (including DMSO, Ethanol, and Isopropanol)
- Suitable for Gamma Irradiation
- Requires 6N for piercing
- Temperature Range: -20°C to +55°C
Autoclavable version:
- Suitable for Gamma Irradiation
- Temperature Range: -20°C to +130°C
Hazardous: No
Storage Condition: Ambient
Specifications
Specifications
| Color | Natural |
| Description | CapMat for 96-well Polypropylene Sample Processing & Storage Plates |
| Certifications/Compliance | Certified RNase, DNase, PCR Inhibitors, and human DNA Free. ANSI Compliant |
| Material | TPE |
| Sterility | Sterile |
| No. of Wells | 96 |
| No. per Case | 50 |
| No. per Pack | 10 |
| Plug Diameter | 7 mm |
| Temperature Range (English) | -4°F to 266°F |
| Show More |