missing translation for 'onlineSavingsMsg'
Learn More

Greiner Bio-One 96-well Sapphire Microplates

Catalog No. 07000313
Change view
Click to view available options
Skirt Style:
Full
Half
Non-skirt
No. per Case:
100
40
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Skirt Style No. per Case
07000313 Half 100
07000105 Full 100
07000107 Half 40
07000312 Non-skirt 100
07000106 Non-skirt 40
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 07000313 Supplier Greiner Bio-One Supplier No. 652290
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Features optimal heat transfer from thermoblock to reaction solution.

  • Free of detectable DNase, RNase, human DNA
  • Non-Pyrogenic
  • Ultra thin polypropylene for optimal heat transfer
  • Are particularly suited for automation
  • Robot-friendly- for grippers, sufficient space for the application

Specifications

Barcode Alphanumeric
Format Standard
Lid Without Lid
Array 8 x 12
Autoclavable Not Autoclavable
Binding Type Non Binding
Color Natural
Product Type PCR Microplate
Skirt Style Half
Material Polypropylene
Volume (Metric) Well 200 μL
No. of Wells 96
Volume (Metric) Working 0.2 mL
Sterility Non-sterile
Surface Treatment Untreated
Well Shape Round, Conical Bottom (V-well)
Dnase Free Yes
For Use With (Application) PCR
No. per Case 100
No. per Pack 10
Plate Compatibility Suitable for ABI
Quantity 100/Cs.
Temperature Range (Metric) -20°C to +60°C
Well Color Clear
Show More Show Less