missing translation for 'onlineSavingsMsg'
Learn More

Greiner Bio-One 96-well Sapphire Microplates

Catalog No. 07000105
Change view
Click to view available options
Skirt Style:
Full
Half
Non-skirt
No. per Case:
100
40
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Skirt Style No. per Case
07000105 Full 100
07000313 Half 100
07000107 Half 40
07000312 Non-skirt 100
07000106 Non-skirt 40
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 07000105 Supplier Greiner Bio-One Supplier No. 652270

Features optimal heat transfer from thermoblock to reaction solution.

  • >Free of detectable DNase, RNase, human DNA
  • Non-Pyrogenic
  • Ultra thin polypropylene for optimal heat transfer
  • Are particularly suited for automation
  • Robot-friendly- for grippers, sufficient space for the application

Specifications

Breakapart No
Format Standard
Lid Without Lid
Array 8 x 12
Binding Type Non Binding
Color Natural
For Use With (Equipment) Wide Range
Skirt Style Full
Material Polypropylene
No. of Wells 96
Sterility Non-sterile
Surface Treatment Non-treated
Total Volume (Metric) Well 200 μL
Well Shape Round, Conical Bottom (V-well)
No. per Case 100
No. per Pack 10
Quantity 100/Cs.
Temperature Range (Metric) -20°C to +60°C
Well Color Clear
Show More Show Less