missing translation for 'onlineSavingsMsg'
Learn More

Greiner Bio-One 96-well Polystyrene Flat Bottom Strip Plates, 2 x 8 Strips
GREENER_CHOICE

Suitable for optical measurements. Greiner Bio-One 96 Well Polystyrene Strip Plates, 2 x 8 Strips, F-Bottom are suitable for non polar proteins and peptides.

$1,774.18 - $1,915.36

Specifications

Format Strip Plate
Color White
Material Polystyrene
No. of Wells 6
Volume (Metric) Working 20 to 350 μL
View More Specs

Products 2
Catalog Number Mfr. No. Surface Treatment Price Quantity & Availability  
Catalog Number Mfr. No. Surface Treatment Price Quantity & Availability  
07000839
GREENER_CHOICE
View Documents Promotion Details
Greiner Bio-One
756070
Microlon 200
Case of 100 Each for $1,774.18
Only null left
 
07000840
GREENER_CHOICE
View Documents Promotion Details
Greiner Bio-One
756071
Microlon 600
Case of 100 Each for $1,915.36
Only null left
 
Description

Description

  • Free of detectable DNase, RNase, human DNA
  • 2 x 8 strips
  • Non-Pyrogenic
  • Light beam not deflected by well profile
  • Suitable for transmission measurements
  • Polystyrene surface is more hydrophobic than surface of hi
Specifications

Specifications

Strip Plate
Polystyrene
20 to 350 μL
388 μL
Yes
5
-20°C to +60°C
White
6
Non-sterile
Flat Bottom (F-well)
100
Yes
Clear
Videos
Safety and Handling

Safety and Handling

ShelfLife : 4 Years after month of production

SDS
Documents

Documents