missing translation for 'onlineSavingsMsg'
Learn More

Greiner Bio-One 96-well Polystyrene Flat Bottom Strip Plates, 1 x 8 strips
GREENER_CHOICE

Catalog No. 07000843
Change view
Click to view available options
Surface Treatment:
FLUOTRAC 200
FLUOTRAC 600
LUMITRAC 200
LUMITRAC 600
Microlon 200
Microlon 600
For Use With (Application):
Fluorescence measurements
Luminescence measurements
Transmission measurements
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Surface Treatment For Use With (Application)
07000843 Microlon 200 Transmission measurements
07000844 Microlon 600 Transmission measurements
07000845 LUMITRAC 600 Luminescence measurements
07000846 LUMITRAC 200 Luminescence measurements
07000847 FLUOTRAC 200 Fluorescence measurements
07000848 FLUOTRAC 600 Fluorescence measurements
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 07000843 Supplier Greiner Bio-One Supplier No. 762070

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Treated specifically to increase protein binding. Greiner Bio-One 96 Well Polystyrene Flat Bottom Strip Plates, 1 x 8 strips are suitable for optical measurements.

  • Free of detectable DNase, RNase, human DNA
  • Non-Pyrogenic
  • Light beam not deflected by well profile
  • Flat Bottom
  • Polystyrene

Specifications

Format Strip Plate
Binding Type Medium Binding
Product Type Strip Plate
Material Polystyrene
No. of Wells 6
Volume (Metric) Working 20 to 350 μL
Sterility Non-sterile
Surface Treatment Microlon 200
Total Volume (Metric) Well 388 μL
Well Shape Flat Bottom (F-well)
Dnase Free Yes
For Use With (Application) Transmission measurements
No. per Case 100
No. per Pack 5
RNase Free Yes
Skirt Color White
Temperature Range (Metric) -20°C to +60°C
Well Color Clear
Show More Show Less