missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRAND™ 96-well Plates
Excellent for cell and tissue culture, sample storage, immunological assays and other applications
Supplier: BRAND™ 701330
Description
- Polypropylene plates are autoclavable at 250°F (121°C) and resistant to many chemicals, including phenols, chloroform and DMSO.
- Clear polystyrene plates are available in non-sterile, sterile, and sterile with treated hydrophilic surfaces, in your choice of flat, round, or conical bottoms.
- Plates have raised rims to simplify sealing with adhesive films, and recessed interwell spaces to reduce well cross-contamination. Optional lids have condensation rings.
Greener Choice Claims
- Environmental benefits include:
- Renewable Energy
Specifications
Standard | |
Clear | |
Polypropylene | |
96 | |
Round |
8 x 12 | |
96-well Assay Microplate | |
330 μL | |
Non-sterile | |
Cell and tissue culture, sample storage, immunological assays and other applications |