missing translation for 'onlineSavingsMsg'
Learn More

Greiner Bio-One 96-well Plate Frame for C8 Single-Break Strips
GREENER_CHOICE

Catalog No. 07000787
Change view
Click to view available options
Sterility:
Non-sterile
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sterility
07000787 Non-sterile
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07000787 Supplier Greiner Bio-One Supplier No. 705185

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For use with 96-well microplate. Greiner Bio-One 96-Well Plate Frame for C8 Single-Break Strip holds microplate in place.

  • 1 x 8 Break-apart C-Bottom
  • Flat w/curved edge

Specifications

Barcode Alphanumeric
Format Strip Plate
Array 8 x 12
Color White
Product Type Frame for single-break-strips
Material Polystyrene
No. of Wells 96
Sterility Non-sterile
Surface Treatment Untreated
Well Shape No Bottom