Learn More
Brands 96-WELL PLATE 1.2 ML 50/PK
Catalog No. NC2120129
Encompass
96-well plate, 1.2 mL, round wells, U shape, polypropylene, 27 mm, 50/pk
Catalog No. NC2120129 Supplier Brands Supplier No. 50439309
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
96-well plate, 1.2 mL, round wells, U shape, polypropylene, 27 mm, 50/pk