Learn More
Thermo Scientific™ 96-Well Microtiter™ Microplates
Catalog No. 1424572
Change view
Click to view available options
Material:
Polystyrene
Well Shape:
Conical Bottom (V-well)
Curved Edges, Flat Bottom (C-well)
Round Bottom (U-well)
Volume (Metric) Well:
280 μL
330 μL
Lid:
Without Lid
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material Well Shape Volume (Metric) Well Lid
1424572 Polystyrene Conical Bottom (V-well) 280 μL Without Lid
1424571 Polystyrene Round Bottom (U-well) 280 μL Without Lid
14245145 Polystyrene Curved Edges, Flat Bottom (C-well) 330 μL Without Lid
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 1424572 Supplier Thermo Scientific™ Supplier No. 2605
Only null left
Use these 96-well microplates for micro-volume applications such as storage, sample or reagent transfer, serial dilution, hemagglutination, blood typing and compliment fixation.
Thermo Scientific™ 96-Well Microtiter Microplates are ideal for micro-volume applications such as storage, sample or reagent transfer, serial dilution, hemagglutination, blood typing and compliment fixation.
  • For high-throughput screening and research
  • Clear plates for colorimetric applications
  • Standard footprint, 128 × 86mm
  • Available as both polystyrene square wells and vinyl square wells
  • Vinyl Microtiter microplates are used for complement fixation, agglutination, and blood typing
  • Styrene Microtiter microplates are used for complement fixation, agglutination, microbiology testing, and blood typing
Ordering Information:
Polystyrene microplates are available as 50 units/package. Vinyl microplates are available as 25 or 100 units/package, depending on product.
Specifications
Format Standard
Lid Without Lid
Array 8 x 12
Packaging 50 Cases, 10 Pk.
Product Line Microtiter
Color Clear
Product Type Microplate
Material Polystyrene
Volume (Metric) Well 280 μL
No. of Wells 96
Sterility Non-sterile
Surface Treatment Non-treated
Well Shape Conical Bottom (V-well)
No. per Case 50
No. per Pack 10
Show More Show Less