missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Greiner Bio-One 96-well Medium Binding Standard ELISA Microplates

For diagnostics and immunological research applications. Greiner Bio-One 96-Well Medium Binding Standard ELISA Microplates is free of detectable DNase, RNase, human DNA.
$310.07 - $368.96
Specifications
| Format | Standard |
|---|---|
| Array | 8 x 12 |
| Binding Type | Medium Binding |
| Color | Clear |
| Material | Polystyrene |
| Catalog Number | Mfr. No. | Well Shape | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Well Shape | Price | Quantity & Availability | |||||
07000100
![]() |
Greiner Bio-One
655001 |
Square, Flat Bottom (F-well) |
Case of 40 Each for $310.07
|
|
|||||
07000613
![]() |
Greiner Bio-One
651001 |
Conical Bottom (V-well) |
Case of 40 Each for $368.96
|
|
|||||
Description
- Non-Pyrogenic
- Free of detectable DNase, RNase, human DNA
- Material: Crystal clear polystyrene
- Alphanumeric well coding
- Suitable for transmission measurements
- Well-to-well spacing: 9mm
- Suitable for tra
Certifications
DIN ISO 9001
Specifications
| Standard | |
| Medium Binding | |
| Polystyrene | |
| Non-sterile | |
| 40/Cs. |
| 8 x 12 | |
| Clear | |
| 96 | |
| Microlon 200 |