Learn More
Greiner Bio-One 96-well CrystalQuick™ Plus Crystallisation Microplates
For automatised applications. Greiner Bio-One 96-Well CrystalQuick™ Plus Crystallisation Microplates are optimised for sealing with VIEWseal adhesive film.
Supplier: Greiner Bio-One 609180
Description
- Non-sterile
- Polystyrene
- Good optical properties
- Hydrophobic surface
- With crystallisation well per reservoir
- Alphanumeric well coding
- With CrystalDrop™ lid, simultaneous experiments are possible with the sitting drop and hanging drop method
Chemical Identifiers
| Standard | |
| Clear | |
| 96 | |
| Square, Flat Bottom (F-well) |
| 8 x 12 | |
| Polystyrene | |
| Non-sterile | |
| Sitting drop applications with different well profiles and material properties |
Specifications
| Standard | |
| Clear | |
| 3.9 μL | |
| Non-sterile | |
| Sitting drop applications with different well profiles and material properties | |
| 80/Cs. |
| 8 x 12 | |
| Polystyrene | |
| 96 | |
| Square, Flat Bottom (F-well) | |
| 80 | |
| 3.9 mm |