missing translation for 'onlineSavingsMsg'
Learn More

Greiner Bio-One 96-well CrystalQuick™ Plus Crystallisation Microplates

Catalog No. 07000561
Change view
Click to view available options
Volume (Metric) Well:
3.9 μL
4.1 μL
Surface Treatment:
Hydrophobic
Hydrophobic LBR
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Well Surface Treatment
07000561 4.1 μL Hydrophobic
07000564 3.9 μL Hydrophobic
07000567 4.1 μL Hydrophobic LBR
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 07000561 Supplier Greiner Bio-One Supplier No. 609130

For automatised applications. Greiner Bio-One 96-Well CrystalQuick™ Plus Crystallisation Microplates are optimised for sealing with VIEWseal adhesive film.

  • Non-sterile
  • Polystyrene
  • Good optical properties
  • Hydrophobic surface
  • With crystallisation well per reservoir
  • Alphanumeric well coding
  • With CrystalDrop™ lid, simultaneous experiments are possible with the sitting drop and hanging drop method

Specifications

Format Standard
Array 8 x 12
Color Clear
Material Polystyrene
Volume (Metric) Well 4.1 μL
No. of Wells 96
Sterility Non-sterile
Surface Treatment Hydrophobic
Well Shape Square, Flat Bottom (F-well)
For Use With (Application) Sitting drop applications with different well profiles and material properties
No. per Case 40
Quantity 40/Cs.
Height (Metric) 14.4 mm
Show More Show Less