Learn More
Greiner Bio-One 96-well CrystalQuick™ Crystallisation Microplates
For automatised applications. Greiner Bio-One 96-Well CrystalQuick™ Crystallisation Microplates are optimised for sealing with VIEWseal adhesive film.
Supplier:  Greiner Bio-One 609801
 Afficher plus de versions de ce produit
Numéro de catalogue. 07000565
Il en reste null
Description
Description
  • Non-sterile
  • Polystyrene
  • Good optical properties
  • With crystallisation well per reservoir
  • Alphanumeric well coding
  • With CrystalDrop™ lid, experiments are possible with sitting drop and hanging drop metho
Spécifications
Identifiants chimiques
Standard
Clear
96
Sitting drop applications with different well profiles and material properties
8 x 12
Polystyrene
Non-sterile
Spécifications
Standard
Inner bags of 10
LBR (low birefringence)
4.1 μL
Non-sterile
Sitting drop applications with different well profiles and material properties
40/Cs.
8 x 12
Clear
Polystyrene
96
Square, Flat Bottom (F-well)
40
14.4 mm
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS