missing translation for 'onlineSavingsMsg'
Learn More

Greiner Bio-One 96-well CrystalQuick™ Crystallisation Microplates

Catalog No. 07000559
Change view
Click to view available options
Well Shape:
Round
Square, Flat Bottom (F-well)
Volume (Metric) Well:
1.9 μL
3.9 μL
4.1 μL
Product Type:
LBR
LBR (low birefringence)
Standard
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Well Shape Volume (Metric) Well Product Type
07000559 Square, Flat Bottom (F-well) 4.1 μL Standard
07000560 Round 1.9 μL Standard
07000563 Square, Flat Bottom (F-well) 3.9 μL Standard
07000565 Square, Flat Bottom (F-well) 4.1 μL LBR (low birefringence)
07000566 Round 1.9 μL LBR
07000568 Square, Flat Bottom (F-well) 3.9 μL LBR
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 07000559 Supplier Greiner Bio-One Supplier No. 609101

For automatised applications. Greiner Bio-One 96-Well CrystalQuick™ Crystallisation Microplates are optimised for sealing with VIEWseal adhesive film.

  • Non-sterile
  • Polystyrene
  • Good optical properties
  • With crystallisation well per reservoir
  • Alphanumeric well coding
  • With CrystalDrop™ lid, experiments are possible with sitting drop and hanging drop metho

Specifications

Format Standard
Array 8 x 12
Color Clear
Product Type Standard
Material Polystyrene
Volume (Metric) Well 4.1 μL
No. of Wells 96
Sterility Non-sterile
Well Shape Square, Flat Bottom (F-well)
For Use With (Application) Sitting drop applications with different well profiles and material properties
No. per Case 40
Quantity 40/Cs.
Height (Metric) 14.4 mm
Show More Show Less