missing translation for 'onlineSavingsMsg'
Learn More

Greiner Bio-One 96-well Conical Bottom (V) Polypropylene Microplates
GREENER_CHOICE

Catalog No. 07000156
Change view
Click to view available options
Color:
Black
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
07000156 Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07000156 Supplier Greiner Bio-One Supplier No. 651209

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

To significantly improve data quality and sensitivity in biochemical assays. Greiner Bio-One 96-Well Conical Bottom (V) Polypropylene Microplates is free of detectable DNase, RNase, human DNA.

  • Uniform external dimensions
  • Well to well spacing of 9mm
  • Features alphanumeric coding
  • Sealable with adhesive films and heat seal

Specifications

Barcode Alphanumeric
Certifications/Compliance Free of detectable DNase/RNase, human DNA and pyrogens.
Format Standard
Lid Without Lid
Array 8 x 12
Autoclavable Not Autoclavable
Color Black
Product Type Storage Microplate
Material Polypropylene
No. of Wells 96
Volume (Metric) Working 50 to 335 μL
Sterility Non-sterile
Surface Treatment Untreated
Well Shape Conical Bottom (V-well)
Max. RCF 4800 x g (swinging bucket rotor)
No. per Case 100
No. per Pack 10
Quantity 100/Cs.
Temperature Range (Metric) -196°C to +121°C
Well Design Chimney
Dimensions (L x W x H) 128 x 85 x 15 mm
Show More Show Less