Learn More
Greiner Bio-One 96-well Chimney Designed No-Bottom Microplates
Description
- Virgin polystyrene construction
- Medium binding surface
- Designed without bottom
- Features well diameter design that offers one-half the well surface area of standard plates
- Allows 50% sample volume reduction without m
Specifications
Specifications
| Array | 8 x 12 |
| Color | Black |
| Material | Polystyrene |
| No. of Wells | 96 |
| Well Design | Chimney |