missing translation for 'onlineSavingsMsg'
Learn More

Greiner Bio-One 96-well, Cell Culture-Treated, Flat-Bottom Microplate

Color:Black, Clear Bottom; Sterility:Sterile; Lid:With Lid

Supplier:  Greiner Bio-One 655866

Numéro de catalogue. 07000630


Il en reste null
Découvrez plus d'offres spéciales

Description

Description

  • Features black pigmented frame with 190μm ultraclear film bottom for ideal compatibility with instrument optics
  • Universal microscope objective compatibility
  • Refractive index similar to glass (1.53)
  • Chimney well design minimizes risk of cross contamination

Service Plan

No

Warranty

No

Spécifications

Spécifications

Standard
8 x 12
Black, Clear Bottom
96
Cell Culture
16
Clear
With Lid
1/Pk., 16/Cs.
Cycloolefin
Sterile
Flat Bottom (F-well)
16/Cs.
Chimney
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
FDS
Documents

Documents

Promotions

Promotions