missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Greiner Bio-One 96-well, Advanced TC Treated μClear™ F-Bottom/Chimney Well Microplates

Description
Advanced TC polymer modification changes plastic surface in a way that increases primary and long-term cell adhesion and positively influences cell-dependant features and function enhanced cell attachment and higher proliferation rates facilitate rapid cell expansion even under restricted growth conditions.
- Provide conditions for improved cell adherence, accelerated cell expansion and increase overall cell yield
- For use with serum reduced and serum-free media
- Allows differentiation of semi-adherent cells
- Non-pyrogenic and non-cytotoxic
- Homogenous cell growth
- Higher transgene activity
- μClear coated well bottom
Specifications
Specifications
| Breakapart | No |
| Certifications/Compliance | Free of detectable DNase/RNase, human DNA and pyrogens; Contents non-cytotoxic |
| Format | Standard |
| Lid | With Lid |
| Culture Area | 34 mm2 |
| Array | 8 x 12 |
| Autoclavable | Not Autoclavable |
| Color | Clear, White |
| Material | Polystyrene |
| No. of Wells | 96 |
| Show More |