Learn More
Sarstedt Inc Micro test plate, 96 well, slip-on lid, flat base, PS, transparent
Catalog No. 501692593
Encompass
Microtest plate 96-well | polystyrene | sterile | non-pyrogenic/endotoxin-free | non-cytotoxic | DNase-/RNase-/DNA-free | labelling: shelf life and batch No. |comes with lid
Catalog No. 501692593 Supplier Sarstedt Inc Supplier No. SRT82.1581.001
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Microtest plate 96-well | polystyrene | sterile | non-pyrogenic/endotoxin-free | non-cytotoxic | DNase-/RNase-/DNA-free | labelling: shelf life and batch No. |comes with lid