Learn More
USA Scientific Cryo-Babies, 1.28 inch x 0.5 inch, white, on rolls
Catalog No. NC9280335
Encompass
Cryo-Babies, large, white labels for 1.5 mL and 2.0 mL tubes, 1.28 inch L x 0.5 inch W, 1000/roll. For -196C to +150C.
Catalog No. NC9280335 Supplier USA Scientific Supplier No. 91882000
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Cryo-Babies, large, white labels for 1.5 mL and 2.0 mL tubes, 1.28 inch L x 0.5 inch W, 1000/roll. For -196C to +150C.