missing translation for 'onlineSavingsMsg'
Learn More

9-Vinylcarbazole 98.0+%, TCI America™

Catalog No. V002125G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
V002125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. V002125G Supplier TCI America Supplier No. V002125G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1484-13-5
Molecular Formula C14H11N
Molecular Weight (g/mol) 193.25
MDL Number MFCD00004966,MFCD00134336
InChI Key KKFHAJHLJHVUDM-UHFFFAOYSA-N
Synonym 9-vinylcarbazole, n-vinylcarbazole, 9-vinyl-9h-carbazole, 9h-carbazole, 9-ethenyl, n-vinyl carbazole, vinylcarbazole, 1-vinylcarbazole, poly 9-vinylcarbazole, carbazole, 9-vinyl, poly-n-vinylcarbazole
PubChem CID 15143
IUPAC Name 9-ethenyl-9H-carbazole
SMILES C=CN1C2=C(C=CC=C2)C2=C1C=CC=C2

Specifications

Melting Point 65°C
Color White-Yellow
Boiling Point 180°C
Quantity 25 g
UN Number 2811
Formula Weight 193.25
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 9-Vinylcarbazole
EINECSNumber (5)-0152
RTECSNumber FE6350000
TSCA Yes