missing translation for 'onlineSavingsMsg'
Learn More

9-Phenylcarbazole, 99%

Catalog No. AAH5682706
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5682706 5 g
AAH5682714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5682706 Supplier Thermo Scientific Chemicals Supplier No. H5682706
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1150-62-5 | C18H13N | 243.31 g/mol

9-Phenylcarbazole has been used in the electrochemical synthesis of poly(9-phenylcarbazole) films via direct anodic oxidation in mixed electrolytes of boron trifluoride diethyl etherate and sulfuric acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
9-Phenylcarbazole has been used in the electrochemical synthesis of poly(9-phenylcarbazole) films via direct anodic oxidation in mixed electrolytes of boron trifluoride diethyl etherate and sulfuric acid.

Solubility
Slightly soluble in water.

Notes
Store away from air and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.

Chemical Identifiers

CAS 1150-62-5
Molecular Formula C18H13N
Molecular Weight (g/mol) 243.31
MDL Number MFCD00004965
InChI Key VIJYEGDOKCKUOL-UHFFFAOYSA-N
PubChem CID 70851
SMILES C1=CC=C(C=C1)N1C2=C(C=CC=C2)C2=C1C=CC=C2

Specifications

Melting Point 95°C to 97°C
Quantity 5 g
Synonym n-phenylcarbazole, 9-phenyl-9h-carbazole, 9h-carbazole, 9-phenyl, carbazole, 9-phenyl, carbazol, 9-phenyl, 9-phenyl carbazole, pubchem23849, 9h-carbazole,9-phenyl, 9-phenylcarbazole, acmc-2099pd
Solubility Information Slightly soluble in water.
IUPAC Name 9-phenylcarbazole
Formula Weight 243.3
Percent Purity 99%
Chemical Name or Material 9-Phenylcarbazole
Hazard Category H315-H318-H335
Hazard Statement GHS H Statement
H318-H335-H315
Causes serious eye damage.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P310-P312-P332+P313-P362-P501c
EINECSNumber 214-564-2
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only