missing translation for 'onlineSavingsMsg'
Learn More
Learn More
9-Phenylcarbazole-3-boronic Acid (contains varying amounts of Anhydride), TCI America™
$298.25 - $298.25
Specifications
| CAS | 854952-58-2 |
|---|---|
| Color | White-Yellow |
| Molecular Formula | C18H14BNO2 |
| MDL Number | MFCD12196936 |
| InChI Key | JWJQEUDGBZMPAX-UHFFFAOYSA-N |
Specifications
| 854952-58-2 | |
| C18H14BNO2 | |
| JWJQEUDGBZMPAX-UHFFFAOYSA-N | |
| 287.13 |
| White-Yellow | |
| MFCD12196936 | |
| 51358450 | |
| Crystalline Powder |