missing translation for 'onlineSavingsMsg'
Learn More

9-Phenylcarbazole-3-boronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. P20015G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P20015G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P20015G Supplier TCI America Supplier No. P20015G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Chemical Name or Material 9-Phenylcarbazole-3-boronic Acid (contains varying amounts of Anhydride)
CAS 854952-58-2
Color White-Yellow
Molecular Formula C18H14BNO2
Quantity 5 g
MDL Number MFCD12196936
Synonym 9-phenyl-9h-carbazol-3-yl boronic acid, 9-phenyl-9h-carbazol-3-ylboronic acid, 9-phenyl carbazole-3-boronic acid, 9-phenyl-3-carbazoleboronic acid, 9-phenylcarbazole-3-boronic acid, b-9-phenyl-9h-carbazol-3-yl boronic acid, n-phenylcarbazole-3-boronic acid, 3-borono-9-phenyl-9h-carbazole, pubchem23232, g00061-watson-int
InChI Key JWJQEUDGBZMPAX-UHFFFAOYSA-N
SMILES OB(O)C1=CC2=C(C=C1)N(C1=C2C=CC=C1)C1=CC=CC=C1
IUPAC Name (9-phenyl-9H-carbazol-3-yl)boronic acid
Molecular Weight (g/mol) 287.13
PubChem CID 51358450
Formula Weight 287.13
Physical Form Crystalline Powder
Show More Show Less