missing translation for 'onlineSavingsMsg'
Learn More
Learn More
9-Phenylcarbazole-3-boronic acid, 98%
Description
9-Phenylcarbazole-3-boronic acid is used in the synthesis of alkyl-functionalized organic dyes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
9-Phenylcarbazole-3-boronic acid is used in the synthesis of alkyl-functionalized organic dyes.
Solubility
Slightly soluble in water.
Notes
Refrigerate. Keep container tightly closed. Store away from oxidizing agent.
9-Phenylcarbazole-3-boronic acid is used in the synthesis of alkyl-functionalized organic dyes.
Solubility
Slightly soluble in water.
Notes
Refrigerate. Keep container tightly closed. Store away from oxidizing agent.
Specifications
Specifications
| Quantity | 1 g |
| Beilstein | 10566519 |
| Solubility Information | Slightly soluble in water. |
| Formula Weight | 287.13 |
| Percent Purity | 98% |
| Physical Form | Solid |
| Chemical Name or Material | 9-Phenylcarbazole-3-boronic acid |
Safety and Handling
| Hazard Category | H315-H319-H335 |
| Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| TSCA | No |
| storageNote1 | 2° to 8°C |
| Recommended Storage | Keep cold; Store under Nitrogen |
RUO – Research Use Only