missing translation for 'onlineSavingsMsg'
Learn More

9-Phenylcarbazole-2-boronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. p-7135438
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P2169200MG 200 mg
P21691G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P2169200MG Supplier TCI America Supplier No. P2169200MG
Only null left

Chemical Identifiers

CAS 1001911-63-2
Molecular Formula C18H14BNO2
Molecular Weight (g/mol) 287.125
MDL Number MFCD22207050
InChI Key XSAOVBUSKVZIBE-UHFFFAOYSA-N
PubChem CID 67286243
IUPAC Name (9-phenylcarbazol-2-yl)boronic acid
SMILES B(C1=CC2=C(C=C1)C3=CC=CC=C3N2C4=CC=CC=C4)(O)O

Specifications

Color White-Yellow
Quantity 200 mg
Formula Weight 287.13
Physical Form Crystalline Powder
Chemical Name or Material 9-Phenylcarbazole-2-boronic Acid (contains varying amounts of Anhydride)