missing translation for 'onlineSavingsMsg'
Learn More

9-Phenylacridine 98.0+%, TCI America™

Catalog No. P119810G
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P119810G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P119810G Supplier TCI America Supplier No. P119810G
Only null left

Chemical Identifiers

CAS 602-56-2
Molecular Formula C19H13N
Molecular Weight (g/mol) 255.32
MDL Number MFCD00092562
InChI Key MTRFEWTWIPAXLG-UHFFFAOYSA-N
PubChem CID 69041
IUPAC Name 9-phenylacridine
SMILES C1=CC=C(C=C1)C2=C3C=CC=CC3=NC4=CC=CC=C42

Specifications

Melting Point 184°C
Quantity 10 g
Formula Weight 255.32
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 9-Phenylacridine