missing translation for 'onlineSavingsMsg'
Learn More

9-Phenanthrenecarboxaldehyde, Thermo Scientific Chemicals

Catalog No. p-3593359
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC130110050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC130110050 Supplier Thermo Scientific Chemicals Supplier No. 130110050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4707-71-5
Molecular Formula C15H10O
Molecular Weight (g/mol) 206.24
MDL Number MFCD00001175
InChI Key QECIGCMPORCORE-UHFFFAOYSA-N
Synonym 9-phenanthrenecarboxaldehyde, phenanthrene-9-carboxaldehyde, 9-formylphenanthrene, phenanthrene-9-aldehyde, 9-phenanthrenecarbaldehyde, 9-phenanthrenealdehyde, 9-phenanthrencarbaldehyd, 9-phenanthraldehyde, 9-phenanthrenaldehyde, phenanthren-9-aldehyd
PubChem CID 78437
IUPAC Name phenanthrene-9-carbaldehyde
SMILES O=CC1=CC2=CC=CC=C2C2=CC=CC=C12

Specifications

Melting Point 100°C
Color Yellow
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 5 g
Formula Weight 206.24
Percent Purity ≥96.0%
Physical Form Powder
Chemical Name or Material 9-Phenanthrenecarboxaldehyde
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 225-194-6

RUO – Research Use Only