missing translation for 'onlineSavingsMsg'
Learn More
Learn More
9-Phenanthreneboronic Acid (contains varying amounts of Anhydride), TCI America™
$42.87 - $42.87
Chemical Identifiers
| CAS | 68572-87-2 |
|---|---|
| Molecular Formula | C14H11BO2 |
| Molecular Weight (g/mol) | 222.05 |
| MDL Number | MFCD00143524 |
| InChI Key | JCDAUYWOHOLVMH-UHFFFAOYSA-N |
| PubChem CID | 11775704 |
Specifications
| 68572-87-2 | |
| MFCD00143524 | |
| 9-phenanthreneboronic acid, 9-phenanthracenylboronic acid, phenanthrene-9-boronic acid, 9-phenanthrenylboronic acid, 9-phenanthrene boronic acid, 9-phenanthreneboronicacid, 9-phenanthrylboronic acid, boronic acid, 9-phenanthrenyl, phenanthracene-9-boronic acid, phenanthreneboronicacid | |
| OB(O)C1=CC2=CC=CC=C2C2=CC=CC=C12 | |
| 222.05 | |
| 222.05 | |
| 9-Phenanthreneboronic Acid (contains varying amounts of Anhydride) |
| C14H11BO2 | |
| 1 g | |
| JCDAUYWOHOLVMH-UHFFFAOYSA-N | |
| (phenanthren-9-yl)boronic acid | |
| 11775704 | |
| Crystalline Powder |
Safety and Handling
TSCA : No