missing translation for 'onlineSavingsMsg'
Learn More

9-Phenanthreneboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. P10931G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P10931G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P10931G Supplier TCI America Supplier No. P10931G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 68572-87-2
Molecular Formula C14H11BO2
Molecular Weight (g/mol) 222.05
MDL Number MFCD00143524
InChI Key JCDAUYWOHOLVMH-UHFFFAOYSA-N
PubChem CID 11775704

Specifications

Quantity 1 g
Synonym 9-phenanthreneboronic acid, 9-phenanthracenylboronic acid, phenanthrene-9-boronic acid, 9-phenanthrenylboronic acid, 9-phenanthrene boronic acid, 9-phenanthreneboronicacid, 9-phenanthrylboronic acid, boronic acid, 9-phenanthrenyl, phenanthracene-9-boronic acid, phenanthreneboronicacid
SMILES OB(O)C1=CC2=CC=CC=C2C2=CC=CC=C12
IUPAC Name (phenanthren-9-yl)boronic acid
Formula Weight 222.05
Physical Form Crystalline Powder
Chemical Name or Material 9-Phenanthreneboronic Acid (contains varying amounts of Anhydride)