missing translation for 'onlineSavingsMsg'
Learn More
Learn More
9-(p-Tolyl)carbazole 98.0+%, TCI America™
Supplier: TCI America T22271G
Specifications
| 9-(p-Tolyl)carbazole | |
| 107°C | |
| C19H15N | |
| OZVXBXKVMMIGDV-UHFFFAOYSA-N | |
| 9-(4-methylphenyl)carbazole | |
| 15798651 | |
| ≥98.0% (GC) |
| 19264-73-4 | |
| Yellow | |
| 1 g | |
| CC1=CC=C(C=C1)N2C3=CC=CC=C3C4=CC=CC=C42 | |
| 257.336 | |
| 257.34 | |
| Crystalline Powder |
Chemical Identifiers
| 19264-73-4 | |
| 257.336 | |
| 15798651 | |
| CC1=CC=C(C=C1)N2C3=CC=CC=C3C4=CC=CC=C42 |
| C19H15N | |
| OZVXBXKVMMIGDV-UHFFFAOYSA-N | |
| 9-(4-methylphenyl)carbazole |
Safety and Handling
TSCA : No